Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134431.2 | NP_001127903.1 | 2 | 878 | ||
NM_130459.3 | NP_569726.2 | 3 | 1144 | |||
GenBank mRNA | AK027677.1 | - | 3 | 1079 | ||
AK055034.1 | - | 2 | 823 | |||
AK092388.1 | - | 1 | 2395 | |||
BC022985.1 | - | 1 | 38 | |||
DB168410.1 | - | 1 | 556 |