Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159587.2 | NP_001153059.1 | 4 | 472 | ||
NM_133493.4 | NP_598000.2 | 4 | 472 | |||
XM_005248659.3 | XP_005248716.1 | 5 | 619 | |||
XM_011535473.1 | XP_011533775.1 | 4 | 317 | |||
XM_017010283.1 | XP_016865772.1 | 2 | 84 | |||
GenBank mRNA | AF410459.1 | - | 4 | 453 | ||
AK095888.1 | - | 4 | 406 | |||
AK313636.1 | - | 4 | 452 | |||
AY149920.1 | - | 4 | 341 | |||
AY788891.1 | - | 4 | 366 | |||
EF553520.1 | - | 4 | 371 |