Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006098.4 | NP_006089.1 | 2 | 254 | ||
GenBank mRNA | AK095666.1 | - | 2 | 252 | ||
AK222488.1 | - | 2 | 254 | |||
AK301029.1 | - | 2 | 90 | |||
AK301468.1 | - | 2 | 250 | |||
AK303316.1 | - | 2 | 262 | |||
AK310916.1 | - | 2 | 264 | |||
AY336089.1 | - | 2 | 151 | |||
BC000214.1 | - | 2 | 272 | 2959178 | ||
BC000366.2 | - | 2 | 242 | 2819249 | ||
BC010119.2 | - | 2 | 229 | 3835636 | ||
BC014256.1 | - | 2 | 231 | 4765908 | ||
BC014788.1 | - | 2 | 238 | 3455986 | ||
BC017287.2 | - | 2 | 219 | 4892271 | ||
BC019093.2 | - | 2 | 232 | 4893346 | ||
BC019362.1 | - | 2 | 236 | 2823761 | ||
BC021993.1 | - | 2 | 254 | 4083071 | ||
BC029996.1 | - | 2 | 81 | |||
BC032006.1 | - | 2 | 233 | 4750184 | ||
BC035460.1 | - | 2 | 235 | |||
CR456978.1 | - | 2 | 148 | |||
CR541909.1 | - | 2 | 148 | |||
GU727632.1 | - | 2 | 148 | |||
M24194.1 | - | 2 | 243 |