Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138283.1 | NP_612140.1 | 4 | 693 | ||
GenBank mRNA | AY707749.1 | - | 4 | 693 | ||
BC117396.1 | - | 3 | 493 | 40125947 |