Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287584.1 | NP_001274513.1 | 10 | 1335 | ||
NM_001287585.1 | NP_001274514.1 | 10 | 1355 | |||
NM_138285.4 | NP_612142.2 | 9 | 1232 | |||
NR_109856.1 | 8 | 1090 | ||||
XM_006712254.2 | XP_006712317.1 | 9 | 1197 | |||
XM_011510576.2 | XP_011508878.1 | 11 | 1463 | |||
XM_011510577.2 | XP_011508879.1 | 11 | 1650 | |||
XM_017003308.1 | XP_016858797.1 | 9 | 1388 | |||
GenBank mRNA | AF411516.1 | - | 9 | 1234 | ||
AF514993.1 | - | 9 | 1190 | |||
AK290760.1 | - | 9 | 1144 | |||
AK298199.1 | - | 10 | 1265 | |||
AK302350.1 | - | 10 | 1304 | |||
BC035597.1 | - | 9 | 1156 | |||
BC047029.1 | - | 9 | 1219 | 5541023 | ||
BC061698.1 | - | 9 | 1186 | 4132440 |