Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138443.3 | NP_612452.1 | 2 | 256 | ||
NR_026978.1 | 2 | 256 | ||||
GenBank mRNA | AK097403.1 | - | 2 | 237 | ||
AK315605.1 | - | 2 | 202 | |||
AY360137.1 | - | 2 | 256 | |||
BC005958.1 | - | 2 | 192 | 4047707 | ||
BC014003.2 | - | 2 | 229 | 3844806 |