Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138785.3 | NP_620140.1 | 2, 3 | 211 | ||
GenBank mRNA | AK289402.1 | - | 2, 3 | 198 | ||
AK308844.1 | - | 2, 3 | 198 | |||
AK308994.1 | - | 19 | ||||
AK315729.1 | - | 2, 3 | 208 | |||
AY358952.1 | - | 2, 3 | 329 | |||
BC014320.1 | - | 2, 3 | 215 | 3686673 |