Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138421.2 | NP_612430.2 | 10 | 1195 | ||
GenBank mRNA | AB489136.1 | - | 10 | 1145 | ||
AK123457.1 | - | 10 | 1186 | |||
BC012010.1 | - | 10 | 1177 | 4473003 |