Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138771.3 | NP_620126.2 | 4 | 1011 | ||
XM_017012775.1 | XP_016868264.1 | 5 | 1251 | |||
XM_017012776.1 | XP_016868265.1 | 4 | 994 | |||
GenBank mRNA | AK026684.1 | - | 2 | 508 | ||
AK289911.1 | - | 4 | 987 | |||
AY358713.1 | - | 4 | 978 | |||
BC012427.1 | - | 2 | 592 | 3858521 |