Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308174.1 | NP_001295103.1 | 11 | 1991 | ||
NM_138790.3 | NP_620145.2 | 11 | 1953 | |||
GenBank mRNA | AK295352.1 | - | 11 | 1946 | ||
AK296188.1 | - | 11 | 1991 | |||
AY358843.1 | - | 11 | 1919 | |||
BC015003.1 | - | 11 | 1882 | 4853026 |