Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138961.2 | NP_620411.2 | 2, 3 | 421 | ||
GenBank mRNA | AF361746.1 | - | 2, 3 | 386 | ||
AK075396.1 | - | 2, 3 | 384 | |||
AK091023.1 | - | 2, 3 | 1336 | |||
AK314710.1 | - | 2, 3 | 384 | |||
AY358382.1 | - | 2, 3 | 373 | |||
BC016868.1 | - | 2, 3 | 255 | 3850665 | ||
DA257581.1 | - | 2, 3 | 421 |