Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127391.2 | NP_001120863.1 | 2, 3 | 575 | ||
NM_001289993.1 | NP_001276922.1 | 2, 3 | 243 | |||
NM_139163.3 | NP_631902.2 | 2, 3 | 575 | |||
NR_110620.1 | 2, 3 | 575 | ||||
XM_011510606.2 | XP_011508908.1 | 2, 3 | 575 | |||
XM_017003360.1 | XP_016858849.1 | 1, 2 | 158 | |||
XM_017003361.1 | XP_016858850.1 | 1, 2 | 158 | |||
XM_017003362.1 | XP_016858851.1 | 1, 2 | 105 | |||
GenBank mRNA | AB053314.1 | - | 1, 2 | 160 | ||
AK124115.1 | - | 2, 3 | 521 | |||
AK292283.1 | - | 2, 3 | 243 | |||
BC031603.1 | - | 2, 3 | 555 | 5168160 | ||
CD703925.1 | - | 2, 3 | 533 | |||
DB034999.1 | - | 2, 3 | 510 |