Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289082.1 | NP_001276011.1 | 3 | 604 | ||
NM_001289083.1 | NP_001276012.1 | 2 | 430 | |||
NM_001289084.1 | NP_001276013.1 | 2 | 430 | |||
NM_001289085.1 | NP_001276014.1 | 3 | 604 | |||
NM_001289086.1 | NP_001276015.1 | 3 | 604 | |||
NM_001289087.1 | NP_001276016.1 | 2 | 430 | |||
NM_139018.4 | NP_620587.2 | 2 | 430 | |||
NR_110298.1 | 3 | 604 | ||||
XM_017024212.1 | XP_016879701.1 | 2 | 482 | |||
XM_017024213.1 | XP_016879702.1 | 2 | 489 | |||
XM_017024214.1 | XP_016879703.1 | 2 | 488 | |||
XM_017024215.1 | XP_016879704.1 | 2 | 430 | |||
XM_017024216.1 | XP_016879705.1 | 2 | 486 | |||
XM_017024217.1 | XP_016879706.1 | 2 | 484 | |||
GenBank mRNA | AF251706.1 | - | 2 | 302 | ||
AF375480.1 | - | 2 | 360 | |||
AF375481.1 | - | 2 | 360 | |||
AK298752.1 | - | 2 | 371 | |||
AK315165.1 | - | 2 | 353 | |||
AY303545.1 | - | 2 | 320 | |||
AY358545.1 | - | 2 | 377 | |||
BC028199.1 | - | 3 | 582 | 5207520 | ||
BI911496.1 | - | 2 | 326 | |||
BM147210.1 | - | 2 | 400 | |||
BM922663.1 | - | 2 | 299 | |||
BP226373.1 | - | 2 | 353 | |||
DA673169.1 | - | 2 | 432 | |||
DQ153249.1 | - | 2 | 305 |