Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142556.1 | NP_001136028.1 | 2 | 254 | ||
NM_012484.2 | NP_036616.2 | 2 | 254 | |||
NM_012485.2 | NP_036617.2 | 2 | 254 | |||
GenBank mRNA | AF032862.1 | - | 2 | 108 | ||
AK290577.1 | - | 2 | 95 | |||
AK302975.1 | - | 2 | 133 | |||
AK313603.1 | - | 2 | 172 | |||
AY623426.1 | - | 2 | 77 | |||
BC017793.1 | - | 2 | 123 | |||
BC030505.2 | - | 2 | 161 | |||
BC057235.1 | - | 2 | 224 | |||
BC070281.1 | - | 2 | 130 | |||
BC108904.1 | - | 2 | 106 | 40007903 | ||
DB091289.1 | - | 2 | 254 | |||
U29343.1 | - | 2 | 132 |