Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139056.3 | NP_620687.2 | 10 | 1671 | ||
NR_136935.1 | 10 | 1671 | ||||
GenBank mRNA | AB095949.1 | - | 7 | 926 | ||
AJ315734.1 | - | 10 | 1533 | |||
AK122980.1 | - | 8 | 1296 | |||
AK127746.1 | - | 10 | 1671 | |||
DQ266047.1 | - | 10 | 1671 |