Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HEXIM2 SIRNA Assays ›
Gene Symbol:
HEXIM2
Gene Name:
hexamethylene bisacetamide inducible 2
Gene Aliases:
L3
Chromosome Location:
Chr.17: 45160174 - 45170040 on Build GRCh38
UniGene ID:
Hs.56382
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37278
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HEXIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303436.1
NP_001290365.1
3
984
NM_001303437.1
NP_001290366.1
4
1437
NM_001303438.1
NP_001290367.1
3
1289
NM_001303439.1
NP_001290368.1
3
1441
NM_001303440.1
NP_001290369.1
3
401
NM_001303441.1
NP_001290370.1
4
549
NM_001303442.1
NP_001290371.1
3
295
NM_001303443.1
NP_001290372.1
4
443
NM_001303444.1
NP_001290373.1
3
298
NM_144608.2
NP_653209.1
4
1132
XM_006721687.3
XP_006721750.1
3
981
XM_011524306.2
XP_011522608.1
4
453
XM_011524307.2
XP_011522609.2
4
888
XM_011524308.2
XP_011522610.2
4
891
XM_017024167.1
XP_016879656.1
4
566
XM_017024168.1
XP_016879657.1
5
1327
XM_017024169.1
XP_016879658.1
4
1179
XM_017024170.1
XP_016879659.1
4
466
GenBank mRNA
AA223138.1
-
3
321
AK056946.1
-
4
409
AY598322.1
-
2
170
BC003531.1
-
3
329
3609101
BC012474.1
-
2
222
4475599
BC025970.1
-
3
259
4559410
BG696988.1
-
3
240
BI829228.1
-
3
523
BI829844.1
-
4
546
BP209696.1
-
3
385
BU501076.1
-
3
428
CN425800.1
-
4
445
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
snRNA binding
7SK snRNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0