Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA5B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039500.2 | NP_001034589.2 | 13 | 2053 | ||
XM_006710360.3 | XP_006710423.2 | 10 | 1328 | |||
XM_011540683.2 | XP_011538985.1 | 13 | 2015 | |||
XM_011540684.2 | XP_011538986.1 | 13 | 2140 | |||
XM_011540685.2 | XP_011538987.1 | 13 | 2152 | |||
XM_011540688.2 | XP_011538990.1 | 13 | 2015 | |||
XM_011540690.2 | XP_011538992.1 | 12 | 1838 | |||
XM_011540691.2 | XP_011538993.1 | 12 | 1829 | |||
XM_011540692.2 | XP_011538994.1 | 13 | 2009 | |||
XM_011540693.1 | XP_011538995.1 | 13 | 2015 | |||
XM_011540695.1 | XP_011538997.1 | 13 | 2015 | |||
XM_011540697.2 | XP_011538999.1 | 7 | 1386 | |||
XM_011540698.1 | XP_011539000.1 | 13 | 2015 | |||
XM_017000305.1 | XP_016855794.1 | 7 | 1132 | |||
XM_017000306.1 | XP_016855795.1 | 8 | 1293 | |||
GenBank mRNA | AK057346.1 | - | 2 | 136 | ||
AK125833.1 | - | 13 | 2201 | |||
AK295797.1 | - | 13 | 2037 | |||
BC101378.1 | - | 2 | 133 | |||
BC101379.2 | - | 2 | 133 | |||
BC101380.1 | - | 2 | 133 | |||
BC101381.1 | - | 2 | 133 |