Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003157.4 | NP_003148.2 | 2 | 367 | ||
XM_006713310.2 | XP_006713373.1 | 2 | 364 | |||
XM_011534039.2 | XP_011532341.1 | 2 | 364 | |||
XM_011534040.2 | XP_011532342.1 | 2 | 364 | |||
XM_017007085.1 | XP_016862574.1 | 2 | 364 | |||
XM_017007086.1 | XP_016862575.1 | 2 | 364 | |||
GenBank mRNA | AK312420.1 | - | 2 | 355 | ||
BC015515.1 | - | 2 | 331 | |||
BC063044.1 | - | 2 | 357 | 5169184 | ||
DB073127.1 | - | 2 | 367 | |||
EF560744.1 | - | 2 | 363 | |||
KJ592714.1 | - | 2 | 164 | |||
L20321.1 | - | 2 | 342 |