Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098502.1 | NP_001091972.1 | 2, 3 | 317 | ||
NM_144636.2 | NP_653237.1 | 3, 4 | 487 | |||
GenBank mRNA | AK056271.1 | - | 3, 4 | 438 | ||
AK290764.1 | - | 2, 3 | 245 | |||
BC017082.1 | - | 2, 3 | 185 | 3848900 | ||
BC033775.1 | - | 2, 3 | 213 | 4538841 |