Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172309.1 | NP_001165780.1 | 6 | 606 | ||
NM_144573.3 | NP_653174.3 | 7 | 798 | |||
XM_005271322.3 | XP_005271379.1 | 7 | 689 | |||
XM_005271323.3 | XP_005271380.1 | 6 | 647 | |||
XM_005271324.4 | XP_005271381.1 | 6 | 593 | |||
XM_005271325.3 | XP_005271382.1 | 7 | 689 | |||
XM_005271326.3 | XP_005271383.1 | 5 | 482 | |||
GenBank mRNA | AF114264.1 | - | 3 | 228 | ||
AF176780.1 | - | 6 | 477 | |||
AK057954.1 | - | 3 | 206 | |||
AK298565.1 | - | 6 | 575 | |||
AK300728.1 | - | 6 | 689 | |||
AK307487.1 | - | 5 | 482 | |||
AY211912.1 | - | 6 | 609 | |||
BC017827.1 | - | 2 | 90 | |||
BC055084.1 | - | 6 | 533 | |||
BC111395.1 | - | 3 | 187 | 4271572 | ||
BC114444.1 | - | 3 | 204 | 40083839 | ||
BC114445.1 | - | 3 | 204 | 40083847 | ||
BX648546.1 | - | 7 | 686 | |||
DQ464902.1 | - | 7 | 685 | |||
S67069.1 | - | 3 | 180 |