Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFXN5 SIRNA Assays ›
Gene Symbol:
SFXN5
Gene Name:
sideroflexin 5
Gene Aliases:
BBG-TCC
Chromosome Location:
Chr.2: 72942036 - 73072234 on Build GRCh38
UniGene ID:
Hs.368171
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37347
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SFXN5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144579.2
NP_653180.1
11
818
XM_005264648.1
XP_005264705.1
11
701
XM_011533186.2
XP_011531488.1
12
2366
XM_011533187.2
XP_011531489.1
12
2363
XM_011533188.2
XP_011531490.1
11
2221
XM_011533189.1
XP_011531491.1
11
724
XM_011533190.2
XP_011531492.1
12
1271
XM_011533192.2
XP_011531494.1
12
1505
XM_011533194.2
XP_011531496.1
12
2364
XM_011533196.2
XP_011531498.1
11
1450
XM_011533197.1
XP_011531499.1
11
701
XM_017005340.1
XP_016860829.1
11
2218
XM_017005341.1
XP_016860830.1
11
2219
XM_017005342.1
XP_016860831.1
11
724
XM_017005344.1
XP_016860833.1
11
1450
XM_017005345.1
XP_016860834.1
3
1823
XM_017005346.1
XP_016860835.1
11
668
GenBank mRNA
AK289731.1
-
11
707
AK294751.1
-
11
755
AK295637.1
-
11
606
AK307116.1
-
2
171
AK308863.1
-
2
158
AK309091.1
-
2
288
AK315861.1
-
1
44
AY044437.1
-
11
822
BC101311.2
-
11
818
40024811
BC101312.1
-
11
818
40024812
BC101313.2
-
11
818
40024813
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
citrate transport
ion transmembrane transport
iron ion homeostasis
citrate transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0