Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040153.3 | NP_001035243.2 | 4 | 547 | ||
NM_001242868.1 | NP_001229797.1 | 4 | 1056 | |||
NM_001242871.1 | NP_001229800.1 | 3 | 595 | |||
NM_144595.3 | NP_653196.1 | 3 | 511 | |||
XM_011534929.2 | XP_011533231.1 | 5 | 936 | |||
XM_011534930.1 | XP_011533232.1 | 4 | 449 | |||
GenBank mRNA | AK054608.1 | - | 3 | 291 | ||
AK289714.1 | - | 3 | 301 | |||
AK294201.1 | - | 4 | 547 | |||
AK295423.1 | - | 3 | 371 | |||
AL834332.1 | - | 3 | 304 | |||
BC045177.1 | - | 3 | 320 | |||
BC111497.1 | - | 3 | 302 | 6148614 | ||
BG391962.1 | - | 435 | ||||
DA343547.1 | - | 3 | 511 |