Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LETM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199659.2 | NP_001186588.1 | 6 | 912 | ||
NM_001199660.2 | NP_001186589.1 | 6 | 751 | |||
NM_001286787.1 | NP_001273716.1 | 6 | 898 | |||
NM_001286819.1 | NP_001273748.1 | 6 | 983 | |||
NM_144652.3 | NP_653253.1 | 5 | 768 | |||
XM_006716291.3 | XP_006716354.1 | 6 | 873 | |||
XM_011544406.1 | XP_011542708.1 | 6 | 872 | |||
XM_011544407.1 | XP_011542709.1 | 6 | 834 | |||
XM_011544408.2 | XP_011542710.1 | 6 | 3479 | |||
XM_011544409.2 | XP_011542711.1 | 6 | 1152 | |||
XM_011544410.2 | XP_011542712.1 | 3 | 400 | |||
XM_017013051.1 | XP_016868540.1 | 6 | 1215 | |||
XM_017013052.1 | XP_016868541.1 | 6 | 943 | |||
XM_017013053.1 | XP_016868542.1 | 6 | 3289 | |||
XM_017013054.1 | XP_016868543.1 | 6 | 928 | |||
XM_017013055.1 | XP_016868544.1 | 6 | 1071 | |||
XM_017013056.1 | XP_016868545.1 | 6 | 917 | |||
XM_017013057.1 | XP_016868546.1 | 5 | 3333 | |||
GenBank mRNA | AK058138.1 | - | 5 | 707 | ||
AK302317.1 | - | 6 | 851 | |||
AK302474.1 | - | 6 | 723 | |||
AY173945.1 | - | 6 | 843 | |||
BC029541.1 | - | 3 | 336 | 5297908 | ||
BC069830.1 | - | 6 | 832 |