Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37433
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
1103
NM_001242416.1
NP_001229345.1
2
1090
NM_001242417.1
NP_001229346.1
4
1366
NM_001242418.1
NP_001229347.1
3
1309
NM_144574.3
NP_653175.2
3
1273
NM_181291.2
NP_851808.1
3
1273
NM_181308.2
NP_851825.1
2
1090
NR_038354.1
4
1066
XM_006720308.1
XP_006720371.1
2
1082
XM_006720309.1
XP_006720372.1
3
1056
XM_006720310.1
XP_006720373.1
3
1017
XM_006720311.1
XP_006720374.1
2
846
XM_006720313.2
XP_006720376.1
4
1025
XM_006720318.2
XP_006720381.1
3
964
XM_011537335.2
XP_011535637.1
4
1385
XM_011537336.2
XP_011535638.1
4
1385
XM_011537337.2
XP_011535639.1
4
1385
XM_011537338.1
XP_011535640.1
4
1332
XM_011537339.2
XP_011535641.1
4
1385
XM_011537340.2
XP_011535642.1
4
1385
XM_011537341.2
XP_011535643.1
3
1291
XM_011537343.1
XP_011535645.1
5
1360
XM_011537344.2
XP_011535646.1
4
1278
XM_011537345.1
XP_011535647.1
3
1056
XM_011537346.1
XP_011535648.1
3
1082
XM_011537347.1
XP_011535649.1
4
1080
XM_011537353.2
XP_011535655.1
4
1136
XM_011537358.2
XP_011535660.1
3
1041
XM_017021766.1
XP_016877255.1
4
1332
XM_017021767.1
XP_016877256.1
3
1291
XM_017021768.1
XP_016877257.1
3
1306
XM_017021770.1
XP_016877259.1
3
1306
XM_017021771.1
XP_016877260.1
4
1341
XM_017021772.1
XP_016877261.1
4
1094
XM_017021773.1
XP_016877262.1
5
1187
XM_017021774.1
XP_016877263.1
4
1136
XM_017021775.1
XP_016877264.1
4
1142
XM_017021776.1
XP_016877265.1
3
1056
XM_017021777.1
XP_016877266.1
3
1024
XM_017021778.1
XP_016877267.1
4
1136
XM_017021779.1
XP_016877268.1
2
846
XM_017021780.1
XP_016877269.1
4
1257
XM_017021781.1
XP_016877270.1
3
964
XM_017021782.1
XP_016877271.1
3
777
XM_017021783.1
XP_016877272.1
4
1073
XM_017021785.1
XP_016877274.1
4
1332
GenBank mRNA
AB209148.1
-
1
5362
AF327354.1
-
3
1198
AK056330.1
-
3
1015
AK090580.1
-
4
1025
AK090978.1
-
3
1044
AK225852.1
-
2
1049
AK297727.1
-
4
1304
AL133558.1
-
1
510
AL832242.1
-
3
1042
AL832500.1
-
3
1248
BC028387.2
-
3
1232
4823662
BX248274.1
-
2
992
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0