Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040153.3 | NP_001035243.2 | 4 | 597 | ||
NM_001242868.1 | NP_001229797.1 | 4 | 1106 | |||
NM_001242871.1 | NP_001229800.1 | 3 | 645 | |||
NM_144595.3 | NP_653196.1 | 3 | 561 | |||
XM_011534929.2 | XP_011533231.1 | 5 | 986 | |||
XM_011534930.1 | XP_011533232.1 | 4 | 499 | |||
GenBank mRNA | AK054608.1 | - | 3 | 341 | ||
AK289714.1 | - | 3 | 351 | |||
AK294201.1 | - | 4 | 597 | |||
AK295423.1 | - | 3 | 421 | |||
AL834332.1 | - | 3 | 354 | |||
BC045177.1 | - | 3 | 370 | |||
BC111497.1 | - | 3 | 352 | 6148614 | ||
BG391962.1 | - | 485 |