Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001158261.1 | NP_001151733.1 | 2 | 382 | ||
NM_144681.2 | NP_653282.2 | 2 | 382 | |||
GenBank mRNA | AK057296.1 | - | 2 | 382 | ||
BC029224.1 | - | 2 | 333 | 5172741 | ||
CV030241.1 | - | 2 | 155 |