Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099433.1 | NP_001092903.1 | 2, 3 | 611 | ||
NM_001306133.1 | NP_001293062.1 | 2, 3 | 401 | |||
NM_144720.3 | NP_653321.1 | 2, 3 | 611 | |||
XM_017007788.1 | XP_016863277.1 | 2, 3 | 575 | |||
XM_017007789.1 | XP_016863278.1 | 2, 3 | 399 | |||
XM_017007790.1 | XP_016863279.1 | 2, 3 | 551 | |||
XM_017007791.1 | XP_016863280.1 | 2, 3 | 575 | |||
XM_017007792.1 | XP_016863281.1 | 2, 3 | 575 | |||
XM_017007793.1 | XP_016863282.1 | 2, 3 | 575 | |||
XM_017007794.1 | XP_016863283.1 | 2, 3 | 575 | |||
XM_017007795.1 | XP_016863284.1 | 2, 3 | 575 | |||
XM_017007796.1 | XP_016863285.1 | 2, 3 | 575 | |||
GenBank mRNA | AK056126.1 | - | 2, 3 | 575 | ||
AK124718.1 | - | 2, 3 | 399 | |||
AM412307.1 | - | 2, 3 | 575 | |||
AM412308.1 | - | 2, 3 | 575 | |||
AM412309.1 | - | 2, 3 | 575 | |||
AY382340.1 | - | 1, 2 | 125 | |||
BC047075.1 | - | 2, 3 | 331 | 5314069 |