Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040424.2 | NP_001035514.1 | 24 | 3693 | ||
NM_001282934.1 | NP_001269863.1 | 25 | 3753 | |||
NM_022115.4 | NP_071398.3 | 31 | 4680 | |||
NR_104257.1 | 25 | 3701 | ||||
NR_104258.1 | 24 | 3581 | ||||
NR_104260.1 | 24 | 3548 | ||||
NR_135464.1 | 24 | 3689 | ||||
XM_006724039.2 | XP_006724102.1 | 24 | 3535 | |||
XM_006724040.2 | XP_006724103.1 | 23 | 3475 | |||
XM_011529674.1 | XP_011527976.1 | 24 | 3565 | |||
XM_011529675.1 | XP_011527977.1 | 25 | 3609 | |||
XM_011529676.2 | XP_011527978.1 | 25 | 6018 | |||
XM_011529677.2 | XP_011527979.1 | 25 | 3694 | |||
XM_011529678.2 | XP_011527980.1 | 25 | 3738 | |||
XM_011529679.2 | XP_011527981.1 | 25 | 4071 | |||
XM_011529681.2 | XP_011527983.1 | 25 | 4975 | |||
XM_011529683.1 | XP_011527985.1 | 30 | 4396 | |||
XM_017028425.1 | XP_016883914.1 | 24 | 3548 | |||
XM_017028426.1 | XP_016883915.1 | 24 | 5957 | |||
GenBank mRNA | AB051812.1 | - | 24 | 3684 | ||
AB051813.1 | - | 25 | 3744 | |||
AB051814.1 | - | 25 | 3692 | |||
AB051815.1 | - | 24 | 3572 | |||
AF426259.1 | - | 4 | 1141 | |||
AF426260.1 | - | 4 | 1141 | |||
AL109788.1 | - | 1 | 433 | |||
AY063456.1 | - | 6 | 1164 | |||
AY078498.2 | - | 31 | 4680 | |||
BC067102.1 | - | 24 | 3548 | 5925140 |