Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSNARE1 SIRNA Assays ›
Gene Symbol:
TSNARE1
Gene Name:
t-SNARE domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 142212080 - 142403291 on Build GRCh38
UniGene ID:
Hs.370931
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37599
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSNARE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291931.1
NP_001278860.1
5
445
NM_145003.4
NP_659440.2
7
1102
XM_011516912.1
XP_011515214.1
6
956
XM_011516913.2
XP_011515215.1
7
1144
XM_011516914.1
XP_011515216.1
7
1055
XM_011516915.2
XP_011515217.1
8
1141
XM_011516916.1
XP_011515218.1
7
1144
XM_011516917.2
XP_011515219.1
7
981
XM_011516918.1
XP_011515220.1
5
806
XM_011516919.1
XP_011515221.1
6
956
XM_011516920.1
XP_011515222.1
4
299
XM_011516921.1
XP_011515223.1
6
956
XM_011516922.1
XP_011515224.1
6
956
XM_011516923.2
XP_011515225.1
2
1283
XM_017013175.1
XP_016868664.1
6
956
XM_017013176.1
XP_016868665.1
6
956
XM_017013177.1
XP_016868666.1
6
956
XM_017013178.1
XP_016868667.1
6
956
XM_017013179.1
XP_016868668.1
6
956
XM_017013180.1
XP_016868669.1
6
956
XM_017013181.1
XP_016868670.1
6
956
GenBank mRNA
AK055726.1
-
7
1031
BC065226.1
-
5
364
BC113556.1
-
6
956
8322608
BC126343.1
-
6
956
8992059
BC143684.1
-
6
956
9052198
BC143686.1
-
6
956
9052200
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
vesicle fusion
vesicle docking
SNARE binding
SNAP receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-kprhh:80/10.239.160.57:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline