Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199380.1 | NP_001186309.1 | 2 | 272 | ||
NM_001199381.1 | NP_001186310.1 | 2 | 319 | |||
NM_001199382.1 | NP_001186311.1 | 2 | 245 | |||
NM_001199383.1 | NP_001186312.1 | 2 | 396 | |||
NM_144726.2 | NP_653327.1 | 2 | 143 | |||
XM_017009138.1 | XP_016864627.1 | 2 | 591 | |||
GenBank mRNA | AK056513.1 | - | 2 | 143 | ||
AK292854.1 | - | 2 | 385 | |||
AK304228.1 | - | 2 | 231 | |||
AK304435.1 | - | 2 | 319 | |||
AK308394.1 | - | 2 | 272 | |||
AK311433.1 | - | 2 | 257 | |||
CK821118.1 | - | 2 | 411 | |||
DC418555.1 | - | 2 | 246 |