Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144676.3 | NP_653277.2 | 4 | 568 | ||
GenBank mRNA | AY358268.1 | - | 4 | 526 | ||
BC020827.1 | - | 4 | 545 | 4766548 | ||
BI600655.1 | - | 4 | 575 |