Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENKUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145010.3 | NP_659447.1 | 2 | 324 | ||
NR_072992.1 | 2 | 324 | ||||
XM_006717389.1 | XP_006717452.1 | 2 | 320 | |||
XM_011519348.1 | XP_011517650.1 | 2 | 320 | |||
GenBank mRNA | AK095021.1 | - | 2 | 269 | ||
AK292495.1 | - | 2 | 321 | |||
AY454125.1 | - | 2 | 311 | |||
BC026165.2 | - | 2 | 326 | 4837530 |