Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289020.1 | NP_001275949.1 | 15 | 1873 | ||
NM_001289021.1 | NP_001275950.1 | 14 | 1705 | |||
NM_001289024.1 | NP_001275953.1 | 2 | 346 | |||
NM_145027.4 | NP_659464.3 | 15 | 1873 | |||
XM_005248904.4 | XP_005248961.1 | 15 | 1873 | |||
XM_011514357.2 | XP_011512659.1 | 15 | 1798 | |||
XM_011514358.2 | XP_011512660.1 | 15 | 1798 | |||
XM_011514359.2 | XP_011512661.1 | 15 | 1798 | |||
XM_017010427.1 | XP_016865916.1 | 14 | 1763 | |||
XM_017010428.1 | XP_016865917.1 | 10 | 1223 | |||
GenBank mRNA | AK123217.1 | - | 2 | 346 | ||
AK131471.1 | - | 15 | 1873 | |||
AL832634.2 | - | 2 | 109 | |||
BC022074.2 | - | 2 | 287 | |||
BC098418.1 | - | 2 | 305 | |||
BC117307.1 | - | 14 | 1635 | 40125858 | ||
BC143444.1 | - | 15 | 1803 | 9051952 | ||
BC143445.1 | - | 14 | 1635 | 9051953 | ||
BX649045.1 | - | 12 | 1453 | |||
DA167995.1 | - | 2 | 346 |