Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM106A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291586.1 | NP_001278515.1 | 3 | 402 | ||
NM_001291588.1 | NP_001278517.1 | 2 | 212 | |||
NM_145041.3 | NP_659478.1 | 3 | 380 | |||
XM_006721658.2 | XP_006721721.1 | 2 | 207 | |||
XM_011524260.1 | XP_011522562.1 | 3 | 334 | |||
XM_017024113.1 | XP_016879602.1 | 3 | 334 | |||
GenBank mRNA | AK056132.1 | - | 3 | 332 | ||
AK290387.1 | - | 3 | 354 | |||
AK301571.1 | - | 2 | 164 | |||
BC012139.2 | - | 2 | 284 | 4562851 | ||
BC146974.1 | - | 2 | 196 | 9056902 | ||
BC146976.1 | - | 2 | 196 | 9056904 | ||
BC147023.1 | - | 2 | 196 | 9056951 | ||
BC157892.1 | - | 2 | 196 | 9057097 |