Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302453.1 | NP_001289382.1 | 5, 6 | 559 | ||
NM_145045.4 | NP_659482.3 | 5, 6 | 853 | |||
XM_017026241.1 | XP_016881730.1 | 5, 6 | 851 | |||
GenBank mRNA | AK056363.1 | - | 5, 6 | 774 | ||
AK294355.1 | - | 7, 8 | 753 | |||
AK302113.1 | - | 5, 6 | 560 | |||
BC014252.2 | - | 4, 5 | 410 | |||
BC141828.1 | - | 5, 6 | 785 | 40148425 | ||
BC142637.1 | - | 5, 6 | 798 | 40147788 | ||
BC144421.1 | - | 3, 4 | 332 |