Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PACRGL SIRNA Assays ›
Gene Symbol:
PACRGL
Gene Name:
PARK2 coregulated like
Gene Aliases:
C4orf28
Chromosome Location:
Chr.4: 20696282 - 20757766 on Build GRCh38
UniGene ID:
Hs.479298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
37911
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PACRGL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130727.1
NP_001124199.1
3
497
NM_001258345.1
NP_001245274.1
3
497
NM_001258346.1
NP_001245275.1
3
288
NM_001317849.1
NP_001304778.1
3
457
NM_145048.3
NP_659485.1
3
497
NR_047661.1
3
321
NR_133933.1
3
354
XM_011513780.1
XP_011512082.1
5
660
XM_011513781.1
XP_011512083.1
5
479
XM_011513784.1
XP_011512086.1
5
496
XM_011513785.1
XP_011512087.1
4
579
XM_011513786.2
XP_011512088.1
5
660
XM_011513787.1
XP_011512089.1
3
451
XM_011513788.1
XP_011512090.1
4
568
XM_011513791.1
XP_011512093.1
3
411
XM_011513792.1
XP_011512094.1
3
406
XM_011513794.1
XP_011512096.1
4
359
XM_011513795.1
XP_011512097.1
3
281
XM_011513796.1
XP_011512098.1
3
276
XM_011513797.2
XP_011512099.1
3
282
XM_011513798.2
XP_011512100.1
3
218
XM_011513804.1
XP_011512106.1
3
243
XM_011513805.2
XP_011512107.1
3
282
XM_017007747.1
XP_016863236.1
5
496
XM_017007748.1
XP_016863237.1
4
366
XM_017007749.1
XP_016863238.1
3
281
XM_017007750.1
XP_016863239.1
4
568
XM_017007751.1
XP_016863240.1
4
564
XM_017007752.1
XP_016863241.1
3
411
XM_017007753.1
XP_016863242.1
3
281
XM_017007754.1
XP_016863243.1
3
497
XM_017007755.1
XP_016863244.1
3
314
XM_017007756.1
XP_016863245.1
3
307
XM_017007757.1
XP_016863246.1
3
243
XM_017007758.1
XP_016863247.1
3
327
XM_017007759.1
XP_016863248.1
3
275
GenBank mRNA
AK098692.1
-
3
354
AK291544.1
-
3
453
AK294077.1
-
3
243
AK297556.1
-
3
308
AK307092.1
-
3
268
AK307355.1
-
3
276
AK307786.1
-
5
496
AK315482.1
-
3
452
BC023002.2
-
3
244
4992139
BM763154.1
-
3
452
BX344401.2
-
3
392
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0