Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145214.2 | NP_660215.1 | 2 | 716 | ||
XM_011544285.2 | XP_011542587.1 | 2 | 783 | |||
XM_017002412.1 | XP_016857901.1 | 2 | 783 | |||
GenBank mRNA | AF327056.1 | - | 2 | 716 | ||
AK074866.1 | - | 2 | 714 | |||
AK097825.1 | - | 2 | 245 | |||
AK226119.1 | - | 1 | 3796 | |||
AK301859.1 | - | 2 | 175 | |||
AK314539.1 | - | 2 | 699 | |||
BC069227.1 | - | 2 | 707 | 4875080 |