Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145207.2 | NP_660208.2 | 15 | 2489 | ||
XM_005262783.4 | XP_005262840.1 | 15 | 2486 | |||
XM_011531678.2 | XP_011529980.1 | 16 | 2558 | |||
XM_017007825.1 | XP_016863314.1 | 16 | 2557 | |||
XM_017007826.1 | XP_016863315.1 | 16 | 2557 | |||
XM_017007828.1 | XP_016863317.1 | 15 | 2451 | |||
XM_017007829.1 | XP_016863318.1 | 14 | 3181 | |||
GenBank mRNA | AF361489.1 | - | 15 | 2485 | ||
AK091384.1 | - | 15 | 2444 | |||
AL832175.1 | - | 15 | 2482 |