Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GCSAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281836.1 | NP_001268765.1 | 3, 4 | 348 | ||
NM_001281837.1 | NP_001268766.1 | 3, 4 | 329 | |||
NM_001281853.1 | NP_001268782.1 | 4, 5 | 433 | |||
NM_145278.4 | NP_660321.1 | 2, 3 | 233 | |||
XM_017000399.1 | XP_016855888.1 | 3, 4 | 348 | |||
GenBank mRNA | AK123798.1 | - | 2, 3 | 144 | ||
AK307082.1 | - | 3, 4 | 314 | |||
AK311994.1 | - | 2, 3 | 184 | |||
BC024174.1 | - | 2, 3 | 133 | 4109064 | ||
DA118297.1 | - | 4, 5 | 367 | |||
DB095029.1 | - | 3, 4 | 329 | |||
DC310064.1 | - | 3, 4 | 349 |