Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD5 SIRNA Assays ›
Gene Symbol:
ACBD5
Gene Name:
acyl-CoA binding domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.10: 27182624 - 27248674 on Build GRCh38
UniGene ID:
Hs.530597
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
38116
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042473.3
NP_001035938.1
3
321
NM_001271512.3
NP_001258441.1
2
635
NM_145698.4
NP_663736.2
3
446
XM_006717528.3
XP_006717591.3
3
6846
XM_006717530.3
XP_006717593.1
3
445
XM_006717531.2
XP_006717594.1
3
455
XM_006717535.2
XP_006717598.1
3
312
XM_017016884.1
XP_016872373.1
3
6846
XM_017016885.1
XP_016872374.1
3
6846
XM_017016886.1
XP_016872375.1
3
6846
XM_017016887.1
XP_016872376.1
3
376
XM_017016888.1
XP_016872377.1
3
441
XM_017016889.1
XP_016872378.1
3
376
XM_017016890.1
XP_016872379.1
3
6846
XM_017016891.1
XP_016872380.1
3
441
XM_017016892.1
XP_016872381.1
3
362
XM_017016893.1
XP_016872382.1
3
6846
XM_017016894.1
XP_016872383.1
3
6846
XM_017016895.1
XP_016872384.1
3
312
XM_017016896.1
XP_016872385.1
3
313
XM_017016897.1
XP_016872386.1
3
441
XM_017016898.1
XP_016872387.1
3
441
XM_017016899.1
XP_016872388.1
3
441
XM_017016906.1
XP_016872395.1
3
313
GenBank mRNA
AK057469.1
-
3
313
AK295031.1
-
2
272
BC030555.1
-
3
350
5218685
BP200763.1
-
3
446
BX643349.1
-
2
580
DC333545.1
-
3
324
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
macroautophagy
pexophagy
aggrephagy
fatty-acyl-CoA binding
molecular_function
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0