Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145267.2 | NP_660310.2 | 2 | 214 | ||
GenBank mRNA | BC018085.1 | - | 2 | 218 | 4817876 | |
BC104649.1 | - | 2 | 209 | 6713316 | ||
BG703280.1 | - | 2 | 223 | |||
CF597237.1 | - | 204 |