Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145808.3 | NP_665807.1 | 2 | 464 | ||
GenBank mRNA | AF106075.1 | - | 2 | 313 | ||
AK056225.1 | - | 2 | 367 | |||
AL834231.1 | - | 2 | 336 | |||
BC028093.1 | - | 2 | 362 | 3876632 | ||
DA325130.1 | - | 2 | 464 |
Gene Symbol: LUZP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128619.2 | NP_001122091.2 | 2 | 464 | ||
GenBank mRNA | DA325904.1 | - | 2 | 386 |