- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: GCSAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281834.1 | NP_001268763.1 | 5 | 595 | ||
NM_001281835.1 | NP_001268764.1 | 4 | 479 | |||
NM_001281836.1 | NP_001268765.1 | 4 | 363 | |||
NM_001281837.1 | NP_001268766.1 | 4 | 344 | |||
NM_001281838.1 | NP_001268767.1 | 3 | 303 | |||
NM_001281853.1 | NP_001268782.1 | 5 | 448 | |||
NM_145278.4 | NP_660321.1 | 3 | 248 | |||
XM_017000399.1 | XP_016855888.1 | 4 | 363 | |||
GenBank mRNA | AK123798.1 | - | 3 | 159 | ||
AK124520.1 | - | 5 | 538 | |||
AK126682.1 | - | 4 | 417 | |||
AK307081.1 | - | 3 | 284 | |||
AK307082.1 | - | 4 | 329 | |||
AK311994.1 | - | 3 | 199 | |||
BC024174.1 | - | 3 | 148 | 4109064 | ||
DA118297.1 | - | 5 | 382 | |||
DB095029.1 | - | 4 | 344 | |||
DC310064.1 | - | 4 | 364 |