Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286401.1 | NP_001273330.1 | 2, 3 | 1215 | ||
NM_145316.3 | NP_660359.2 | 3, 4 | 782 | |||
NR_104442.1 | 2, 3 | 684 | ||||
NR_104443.1 | 1, 2 | 599 | ||||
XM_011514362.1 | XP_011512664.1 | 4, 5 | 794 | |||
XM_011514364.2 | XP_011512666.1 | 4, 5 | 1578 | |||
XM_011514365.2 | XP_011512667.1 | 4, 5 | 1363 | |||
XM_011514367.2 | XP_011512669.1 | 3, 4 | 709 | |||
XM_011514369.2 | XP_011512671.1 | 1, 2 | 110 | |||
XM_011514370.2 | XP_011512672.1 | 1, 2 | 100 | |||
GenBank mRNA | AK098666.1 | - | 3, 4 | 741 | ||
BC026012.1 | - | 2, 3 | 654 | 5269554 | ||
BC029657.1 | - | 1, 2 | 80 | 5167418 | ||
DB458508.1 | - | 2, 3 | 166 |