Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL9 SIRNA Assays ›
Gene Symbol:
OSBPL9
Gene Name:
oxysterol binding protein like 9
Gene Aliases:
ORP-9, ORP9
Chromosome Location:
Chr.1: 51616874 - 51789219 on Build GRCh38
UniGene ID:
Hs.21938
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
38339
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024586.5
NP_078862.4
14
1388
NM_148904.3
NP_683702.1
10
1105
NM_148905.3
NP_683703.1
9
1009
NM_148906.2
NP_683704.2
13
1337
NM_148907.2
NP_683705.1
10
982
NM_148908.3
NP_683706.3
13
1349
NM_148909.3
NP_683707.3
14
1418
NR_036662.1
13
1309
XM_006710318.2
XP_006710381.1
13
1148
XM_006710320.3
XP_006710383.1
11
985
XM_006710321.2
XP_006710384.1
10
981
XM_006710322.3
XP_006710385.1
11
1142
XM_006710323.2
XP_006710386.1
10
1050
XM_006710324.3
XP_006710387.1
12
1060
XM_006710325.3
XP_006710388.1
11
1030
XM_006710326.2
XP_006710389.1
9
1032
XM_011540599.2
XP_011538901.1
15
1242
XM_011540600.2
XP_011538902.1
14
1209
XM_011540601.2
XP_011538903.1
13
1170
XM_011540602.2
XP_011538904.1
14
1144
XM_011540603.2
XP_011538905.1
14
1168
XM_011540604.2
XP_011538906.1
11
943
XM_017000218.1
XP_016855707.1
12
1109
XM_017000219.1
XP_016855708.1
13
1129
XM_017000220.1
XP_016855709.1
10
870
XM_017000221.1
XP_016855710.1
10
821
XM_017000222.1
XP_016855711.1
14
1354
XM_017000223.1
XP_016855712.1
14
1352
XM_017000224.1
XP_016855713.1
11
1297
XM_017000225.1
XP_016855714.1
12
1148
GenBank mRNA
AB208851.1
-
3
655
AF392445.1
-
14
1162
AK022554.1
-
9
789
AK027535.1
-
13
1119
AK027707.1
-
10
945
AK056617.1
-
14
1207
AK091703.1
-
13
1091
AK124037.1
-
5
1635
AK128043.1
-
16
1612
AK297656.1
-
11
1135
AK315167.1
-
10
981
AY178997.1
-
13
1111
BC025978.2
-
10
1015
3621193
CR457288.1
-
7
656
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline