Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152284.3 | NP_689497.1 | 1 | 281 | ||
GenBank mRNA | AB120734.1 | - | 1 | 113 | ||
AK000049.1 | - | 1 | 244 | |||
AK314873.1 | - | 1 | 280 | |||
AY329086.1 | - | 1 | 115 | |||
BC014321.1 | - | 1 | 285 | 3828410 | ||
BX492886.1 | - | 1 | 301 |