Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152289.2 | NP_689502.2 | 6 | 640 | ||
XM_005260150.2 | XP_005260207.1 | 7 | 778 | |||
XM_005260151.2 | XP_005260208.1 | 5 | 481 | |||
XM_006722938.2 | XP_006723001.1 | 6 | 595 | |||
XM_011528433.2 | XP_011526735.1 | 6 | 859 | |||
XM_011528434.2 | XP_011526736.1 | 7 | 765 | |||
XM_011528436.2 | XP_011526738.1 | 7 | 779 | |||
XM_011528437.2 | XP_011526739.1 | 7 | 770 | |||
XM_017027478.1 | XP_016882967.1 | 7 | 774 | |||
XM_017027479.1 | XP_016882968.1 | 6 | 663 | |||
XM_017027480.1 | XP_016882969.1 | 5 | 519 | |||
XM_017027481.1 | XP_016882970.1 | 5 | 712 | |||
XM_017027482.1 | XP_016882971.1 | 5 | 547 | |||
GenBank mRNA | AK122974.1 | - | 5 | 517 | ||
AK293004.1 | - | 6 | 630 | |||
AK304383.1 | - | 5 | 547 | |||
BC012144.2 | - | 5 | 561 | |||
BC032668.1 | - | 6 | 632 | 5540009 |