Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135218.1 | NP_001128690.1 | 2, 3 | 698 | ||
NM_005644.3 | NP_005635.1 | 2, 3 | 361 | |||
XM_017002186.1 | XP_016857675.1 | 2, 3 | 763 | |||
GenBank mRNA | AK310097.1 | - | 2, 3 | 355 | ||
BC011986.1 | - | 2, 3 | 611 | 3882343 | ||
BE787487.1 | - | 2, 3 | 589 | |||
BT007031.1 | - | 1, 2 | 176 | |||
BX095156.1 | - | 2, 3 | 361 | |||
CX865268.1 | - | 2, 3 | 620 | |||
D50544.1 | - | 1, 2 | 262 | |||
U57693.1 | - | 2, 3 | 301 | |||
X84002.1 | - | 2, 3 | 342 |