Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152310.2 | NP_689523.1 | 1 | 359 | ||
XM_011540245.2 | XP_011538547.1 | 2 | 1349 | |||
GenBank mRNA | BC034344.1 | - | 1 | 251 | 4745607 | |
HY050356.1 | - | 1 | 360 |