Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147193.2 | NP_671726.2 | 8, 9 | 2112 | ||
XM_017000408.1 | XP_016855897.1 | 9, 10 | 2179 | |||
XM_017000409.1 | XP_016855898.1 | 9, 10 | 2167 | |||
XM_017000410.1 | XP_016855899.1 | 9, 10 | 2293 | |||
GenBank mRNA | AK090634.1 | - | 8, 9 | 2111 | ||
AK093474.1 | - | 6, 7 | 1481 | |||
BC101799.1 | - | 6, 7 | 1618 | 8069305 | ||
BC104911.1 | - | 6, 7 | 1618 | 8143914 |